missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Reflectoquant™ Potassium Test Strips

Catalog No. M1169920001
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1169920001 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1169920001 Supplier MilliporeSigma™ Supplier No. 1.16992.0001
Only null left

Includes

Reagent, if needed

Used in reflectometric test

  • To determine potassoim ions in drinking and mineral waters, industrial water, wastewater, beverages as well as soils after pretreatment

Range: 0.25 to 1.2g/L

Order Info

Pack of 50 strips

Specifications

For Use With (Application) The test can be used for the determination of potassium ions in drinking water and mineral water, industrial water, wastewater, beverages and soils after sample pretreatment
Graduations 0.25 to 1.2g/L
Includes 1 barcode strip, 2 bottles of reagent K-1, 2 mini-test tubes
Quantity 50 Tests
Range 0.25 to 1.2 g/L
Storage Requirements 15°C to 25°C
Test Method Reflectometric
Type Reflectoquant™ Potassium Test Strip