missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Reflectoquant™ Peracetic Acid Test Strips

Catalog No. M1169750001
Change view
Click to view available options
Range:
1 to 22.5mg/L
75 to 400mg/L
Graduations:
1 to 22.5mg/L
75 to 400mg/L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range Graduations
M1169750001 1 to 22.5mg/L 1 to 22.5mg/L
M1169760001 75 to 400mg/L 75 to 400mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1169750001 Supplier MilliporeSigma™ Supplier No. 1.16975.0001

Includes

Tube containing 50 strips

For determination of inorganic peracetic acid in aqueous solutions

  • Redox reaction method
  • Barcode calibration for reliable, quantitative results
  • Low analysis costs

Reclose the tube containing the test strips immediately after use.

For food and water analysis

Specifications

For Use With (Application) Food and water analysis, ideal for determining when hydrogen peroxide is present in a sample
For Use With (Equipment) RQflex™ 10 and RQflex™ Plus 10 reflectometers
Graduations 1 to 22.5mg/L
No. of Tests 50
Range 1 to 22.5mg/L
Storage Requirements 2°C to 8°C
Test Method Redox Reaction
Type Test Strip