missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Reflectoquant™ Nitrite Test Strips

Catalog No. M1169730001
Change view
Click to view available options
Range:
0.03 to 1g/L
0.5 to 25mg/L
Test Method:
Aromatic Amine
Griess' reaction
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range Test Method
M1169730001 0.5 to 25mg/L Griess' reaction
M1167320001 0.03 to 1g/L Aromatic Amine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1169730001 Supplier MilliporeSigma™ Supplier No. 1.16973.0001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Tube containing 50 test strips, 1 bar-code strip.

To determine the presence or absence of residues of nitrite

  • Griess's reaction method
  • Barcode calibration for reliable, quantitative results
  • Low analysis costs

Reclose the tube containing the test strips immediately after use.

For food and water analysis

Specifications

For Use With (Application) Ideal for monitoring nitrite levels in drinking and groundwater as well as in-process, wastewater or aquariums, food and water analysis
For Use With (Equipment) RQflex™ 10 and RQflex™ Plus 10 reflectometers
Graduations 0.5 to 25mg/L
No. of Tests 50
Range 0.5 to 25mg/L
Storage Requirements 2°C to 8°C
Test Method Griess' reaction
Type Test Strip