Learn More
MilliporeSigma™ Reflectoquant™ Nitrite Test Strips
To determine the presence or absence of residues of nitrite
Supplier: MilliporeSigma™ 1.16732.0001
Includes: Tube containing 50 test strips, 1 bar-code strip.
Description
- Griess's reaction method
- Barcode calibration for reliable, quantitative results
- Low analysis costs
Reclose the tube containing the test strips immediately after use.
For food and water analysis
Chemical Identifiers
| 0.5 to 25mg/L | |
| 50 | |
| 2°C to 8°C | |
| Test Strip |
Specifications
| For food and water analysis | |
| 0.5 to 25mg/L | |
| 0.03 to 1g/L | |
| Aromatic Amine |
| Reflectometer | |
| 50 | |
| 2°C to 8°C | |
| Test Strip |