missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Reflectoquant™ Nitrate Test Strips

Catalog No. p-4628012
Change view
Click to view available options
Range:
3 to 90mg/L
5 to 225mg/L
Graduations:
3 to 90mg/L
5 to 225mg/L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range Graduations
M1169710001 5 to 225mg/L 5 to 225mg/L
M1169950001 3 to 90mg/L 3 to 90mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1169710001 Supplier MilliporeSigma™ Supplier No. 1.16971.0001
Only null left

Includes

Tube containing 50 test strips, 1 bar-code strip.

To determine the presence or absence of residues of nitrate

  • Modified Griess' reaction method
  • Barcode calibration for reliable, quantitative results
  • Low analysis costs

Reclose the tube containing the test strips immediately after use.

For food and water analysis

Specifications

For Use With (Application) Rapid determination of the nitrate content is of particular interest with fruit and vegetables
For Use With (Equipment) Reflectometer
Graduations 5 to 225mg/L
No. of Tests 50
Range 5 to 225mg/L
Storage Requirements 2°C to 8°C
Test Method Modified Griess' reaction
Type Test Strip