Learn More
MilliporeSigma™ Reflectoquant™ Malic Acid Test Strips
Used for determination of decrease in malic acid concentration
Supplier: MilliporeSigma™ 1161280001
Includes: Tube containing 50 strips, 1 bar-code strip
Description
- Enzymatic reaction method
- Barcode calibration for reliable, quantitative results
- Low analysis costs
For analysis of breverages and food after sample pretreatment
Specifications
| Analysis of beverages and food after sample pretreatment | |
| 5 to 60mg/L | |
| 5 to 60mg/L | |
| Enzymatic Reaction |
| RQflex™ 10 and RQflex™ Plus 10 reflectometers | |
| 50 | |
| 2°C to 8°C | |
| Test Strip |