missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Reflectoquant™ Magnesium Test Strips
Description
Includes
Reagent, if needed
- For determining magnesium ions in goundwater and surface water, drinking water, mineral water, boiler water and food after pretreating
Range: 5 to 100mg/L
Order Info
Pack of 50 strips
Specifications
Specifications
| For Use With (Application) | Used for the determination of magnesium ions in groundwater and surface water, drinking water, mineral water, boiler water and food after pretreatment |
| For Use With (Equipment) | Used with the Reflectoquant™ RQflex™ 10m reflectometer analysis system |
| Graduations | 5 to 100mg/L |
| Includes | Barcode strip |
| Quantity | 50 Tests |
| Storage Requirements | 2°C to 8°C |
| Test Method | Phthaleinkomplexone |
| Type | Magnesium Test Strip |