missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Reflectoquant™ Lactic acid Test Strips
Description
Includes
Reagent, if needed
- Test measures total lactic acid (sum of D- and L-lactic acid) in beverages , e.g., wine, beer, fruit/vegetable juices after dilution
- Can be used to determine lactic acid in food after pretreatment
Range: 3.0 to 60.0mg/L
Order Info
Pack of 50 strips
Specifications
Specifications
| For Use With (Application) | Ideal for the determination of lactic acid total lactic acid in beverages like wine, beer and fruit or vegetable juice after dilution |
| For Use With (Equipment) | Designed for use with the RQflex™ 10 or RQflex plus 10 reflectometer meters |
| Graduations | 3 to 60mg/L |
| Includes | 1 barcode strip |
| Quantity | 50 Tests |
| Range | 3 to 60 mg/L |
| Storage Requirements | 2°C to 8°C |
| Test Method | Reflectometric |
| Type | Reflectoquant™ Lactic Acid Test Strip |