missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Reflectoquant™ Iron Test Strips

Catalog No. M1169820001
Change view
Click to view available options
Range:
0.5 to 20mg/L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
M1169820001 0.5 to 20mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1169820001 Supplier MilliporeSigma™ Supplier No. 1.16982.0001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Tube containing 50 test strips, 1 bar-code strip

To determine the presence of residues of iron

  • Triazine reaction method
  • Barcode calibration for reliable, quantitative results
  • Low analysis costs

Reclose the tube containing the test strips immediately after use.

For food and water analysis

Specifications

For Use With (Application) Ideal for measuring iron(II) ions in surface water, seawater, industrial water, wastewater and mineral water
For Use With (Equipment) RQflex™ 10 and RQflex™ Plus 10 reflectometers
Graduations 0.5 to 20mg/L
No. of Tests 50
Range 0.5 to 20mg/L
Storage Requirements 15°C to 25°C
Test Method Triazine
Type Test Strip