missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Reflectoquant™ Formaldehyde Test Strips

Catalog No. M1169890001
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1169890001 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1169890001 Supplier MilliporeSigma™ Supplier No. 1.16989.0001
Only null left

Includes

Reagent, if needed

Used in reflectometric test

  • For determining formaldehyde in aqueous solutions and in disinfecting and rinsing solutions in laundries

Range: 1.0 to 45.0mg/L

Order Info

Pack of 50 strips

Specifications

For Use With (Application) Used for the determination of formaldehyde in disinfecting and rinsing solutions
For Use With (Equipment) Reflectometric
Graduations 1 to 45mg/L
Includes 1 barcode strip, 2 bottles of reagent Fo-1, 1 test vessel with stopper
Quantity 50 Tests
Range 1 to 45 mg/L
Storage Requirements 15°C to 25°C
Test Method Triazole
Type Formaldehyde Test Strip