missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Reflectoquant™ Chlorine Test Strips

Catalog No. M1168960001
Change view
Click to view available options
pH Range:
5 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
M1168960001 5 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1168960001 Supplier MilliporeSigma™ Supplier No. 1.16896.0001
Only null left

Includes

Reagent, if needed

Used in reflectometric test

  • For determination of chlorine in water disinfection from treatment of drinking and wastewater to pool and spa sanitization

Range: 0.5 to 10mg/L

Order Info

Pack of 50 strips

Specifications

pH Range 5 to 10
For Use With (Application) Used for determination of chlorine in the area of water disinfection, where applications vary from treatment of drinking and wastewater to pool and spa sanitization
Graduations 0.5 to 10mg/L
Includes Barcode strip, Cl2-1 reagent bottle, 1 test vessel with stopper
Quantity 50 Tests
Storage Requirements 2°C to 8°C
Test Method Redox reaction
Type Chlorine Test Strip