missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Reflectoquant™ Calcium Test Kit

Catalog No. M1169930001
Change view
Click to view available options
Graduations:
2.5 to 45mg/L
5 to 125mg/L
Range:
2.5 to 45 mg/L
5 to 125 mg/L
Storage Requirements:
15°C to 25°C
2°C to 8°C
Test Method:
Glyoxalbishydroxyanil
Phthaleinkomplexone
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Graduations Range Storage Requirements Test Method
M1169930001 2.5 to 45mg/L 2.5 to 45 mg/L 15°C to 25°C Glyoxalbishydroxyanil
M1161250001 5 to 125mg/L 5 to 125 mg/L 2°C to 8°C Phthaleinkomplexone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1169930001 Supplier MilliporeSigma™ Supplier No. 1.16993.0001
Only null left

Includes

Reagent, if needed

Used in reflectometric test

  • For testing boiling and cooling water, drinking water, beer
  • Indicates possible presence of calcium deposits that can affect operation of machinery

Range: 5 to 125mg/L

Order Info

Pack of 50 strips

Specifications

For Use With (Equipment) Reflectometric
Graduations 2.5 to 45mg/L
Includes 1 barcode strip, 1 bottle reagent Ca-1, 1 bottle reagent Ca-2, 1 test vessel with stopper
Quantity 50 Tests
Range 2.5 to 45 mg/L
Storage Requirements 15°C to 25°C
Test Method Glyoxalbishydroxyanil
Type Calcium Test Strip