missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Reflectoquant™ Calcium Test Kit
Description
Includes
Reagent, if needed
- For testing boiling and cooling water, drinking water, beer
- Indicates possible presence of calcium deposits that can affect operation of machinery
Range: 5 to 125mg/L
Order Info
Pack of 50 strips
Specifications
Specifications
| For Use With (Equipment) | Reflectometric |
| Graduations | 5 to 125mg/L |
| Quantity | 50 Tests |
| Range | 5 to 125 mg/L |
| Storage Requirements | 2°C to 8°C |
| Test Method | Phthaleinkomplexone |
| Type | Calcium Test Strip |