Learn More
MilliporeSigma™ Reflectoquant™ Blank Test Strips
test strips without reagents Reflectoquant™
Supplier:  MilliporeSigma™ 1.16730.0001
Catalog No. M1167300001
Only null left
Includes: Reagent, if needed
Description
Description
  • Strips without reagents
  • Use to correct false-high measurement values when testing already-colored samples
Order Info
Pack of 50 strips
Specifications
Specifications
Blank strips can be used to correct the measurement values without having to take time-consuming sample-pretreatment measures
50 Tests
15°C to 25°C
Reflectoquant™ Blank Strip
Product Suggestions
Product Suggestions