missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Reflectoquant™ Ascorbic Acid Test Strips

Catalog No. M1169810001
Change view
Click to view available options
Range:
25 to 450mg/L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
M1169810001 25 to 450mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1169810001 Supplier MilliporeSigma™ Supplier No. 1.16981.0001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Tube containing 50 test strips, 1 bar-code strip

To determine the presence of ascorbic acid

  • Phosphormolybdenum blue reaction method
  • Barcode calibration for reliable, quantitative results
  • Low analysis costs

Reclose the tube containing the test strips immediately after use.

For food and water analysis

Specifications

For Use With (Application) Ideal for testing for ascorbic acid in beverages (e.g. fruit and vegetable juices, sodas, wine and beer)
For Use With (Equipment) Reflectoquant™ RQflex™ 10m reflectometer analysis system
Graduations 25 to 450mg/L
No. of Tests 50
Range 25 to 450mg/L
Storage Requirements 15°C to 25°C
Test Method Phosphormolybdenum Blue
Type Test Strip