missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Quantum EX Cartridge

Catalog No. QTUMMPKEX
Change view
Click to view available options
For Use With (Equipment):
Direct-Q™ 5, Milli-Q™ Academic, Biocel, Element, Gradient, Synthesis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
QTUMMPKEX Direct-Q™ 5, Milli-Q™ Academic, Biocel, Element, Gradient, Synthesis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QTUMMPKEX Supplier MilliporeSigma™ Supplier No. QTUMMPKEX

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Nonsterile Millipak™-40 point-of-use final filter

Application-specific for removal of organics

  • Utilizes Organex resin

Specifications

Pore Size 0.2 μm
Purification Method Ion Exchange, Activated Carbon, Membrane Filter
Resin Type Organex
For Use With (Equipment) Direct-Q™ 5, Milli-Q™ Academic, Biocel, Element, Gradient, Synthesis
Model Quantum EX with non-sterile Millipak™-40
Type Cartridge