missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Purification Kit

Catalog No. p-4903946
Change view
Click to view available options
Type:
Purification Cartridge
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
QGARD00D3 Purification Cartridge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QGARD00D3 Supplier MilliporeSigma™ Supplier No. QGARD00D3
Only null left

With ion-exchange technology

Q-Gard™ 3 Purification Cartridge for Milli-Q™ systems connected to DI water feed containing high silt

Specifications

Feedwater Source Deionized
Purification Method Ion Exchange, Activated Carbon, Depth Filtration
For Use With (Equipment) Milli-Q™systems connected to DI water feed containing high silt
Model Q-Gard 3
Type Purification Cartridge