missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Purification Kit
Description
Q-Gard™ 3 Purification Cartridge for Milli-Q™ systems connected to DI water feed containing high silt
Specifications
Specifications
| Feedwater Source | Deionized |
| Purification Method | Ion Exchange, Activated Carbon, Depth Filtration |
| For Use With (Equipment) | Milli-Q™systems connected to DI water feed containing high silt |
| Model | Q-Gard 3 |
| Type | Purification Cartridge |