missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Pump Connection Kit for Pure Water Storage Reservoir Distribution Pump

Catalog No. PUMPKIT01
Change view
Click to view available options
For Use With (Equipment):
Distribution Pump (TANKREC56)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
PUMPKIT01 Distribution Pump (TANKREC56)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PUMPKIT01 Supplier MilliporeSigma™ Supplier No. PUMPKIT01

Pump Connection Kit (PUMP KIT 01) contains 12mm polyethylene tubing, check valve, connectors, elbows and tees.

Contains items needed to connect the TANKREC56 distribution pump.

Specifications

For Use With (Equipment) Distribution Pump (TANKREC56)
Includes 12mm Polyethylene tubing, check valve, connectors, elbows and tees