missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Progard Kit
Description
Progard™ 2 Pack & vent filter for Type 2
Specifications
Specifications
| Purification Method | CaO, Activated Carbon, Anti-scaling, Depth Filtration, Membrane Filter |
| For Use With (Application) | Filters large particles from tap water and reduces hardness prior to reverse osmosis purification |
| For Use With (Equipment) | For Type 2 |
| Includes | 1x Progard 2, one 1x Vent Filter for Type 2 |
| Quantity | 2 Pack |
| Type | Vent Filter |