missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Presenilin-2, Blocking Peptide for AB5056

Catalog No. ag216mi
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AG216MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AG216MI Supplier MilliporeSigma™ Supplier No. AG216
Only null left

Specifications

Gene ID (Entrez) NM_000447.2, NM_012486.2
Concentration 1.0 mg/mL
Content And Storage -20°C to -70°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles
Format Liquid
Formulation Liquid in sterile water. Contains no preservative.
Gene Symbol PSEN2, AD5, AD3L, STM2, STM-2, AD3LP, E5-1, Presenilin-2, PSNL2, PS-2, PS2, AD4, EC 3.4.23.- [Contains: Presenilin-2 NTF subunit, Presenilin-2 CTF subunit].
Product Type Presenilin-2, blocking peptide for AB5056
Quantity 100 μg
Regulatory Status RUO - research use only