missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ PER1 (Hu) Control Peptide

Catalog No. AG330MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AG330MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AG330MI Supplier MilliporeSigma™ Supplier No. AG330

Specifications

Gene ID (Entrez) NM_152831.1, NM_000789.2, NM_152830.1
Concentration 1.0 mg/mL
Content And Storage -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Format Liquid
Formulation Liquid in PBS, pH 7.
Gene Symbol ACE, DCP, ACE1, DCP1, carboxycathepsin, ACE-T, MGC26566, CD143, EC 3.4.15.1, EC 3.2.1.- [Contains: Angiotensin-converting enzyme, testis- specific isoform, soluble form]., EC 3.4.15.1 [Contains: Angiotensin-converting enzyme, somatic isoform, soluble form].
Product Type PER1 (Hu), control peptide for AB5424P
Quantity 100 μg
Regulatory Status RUO