missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Packs and Accessories for Milli-RO™ Compact Water Systems: RO Membrane

Catalog No. CDRC00201
Change view
Click to view available options
RO membrane
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CDRC00201 RO membrane
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDRC00201 Supplier MilliporeSigma™ Supplier No. CDRC00201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

RO membrane for Milli-RO Compact Water Systems

For Elix 3 and RiOs 3 and 5 water purification systems.

  • For removing ionic and organic contaminants down to trace levels from RO, electro-deionised or distilled feed water

Specifications

Feedwater Source Electrically Deionized, Distilled
Purification Method Reverse osmosis
For Use With (Application) For removing ionic and organic contaminants down to trace levels from RO, electro-deionised or distilled feed water
For Use With (Equipment) Elix™, RiOs™ 3, RiOs, Milli-RO™
Model RO
Type RO Membrane