missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ P2Y2 Control Peptide

Catalog No. AG378MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AG378MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AG378MI Supplier MilliporeSigma™ Supplier No. AG378
Only null left

Specifications

Gene ID (Entrez) NM_002564.2, NM_176071.1, NM_176072.1
Concentration 1 mg/mL
Content And Storage -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Format Liquid
Formulation Liquid.
Gene Symbol P2RY2, P2U, MGC40010, P2RU1, P2Y2, P2Y2R, P2U1, P2UR, MGC20088, HP2U
Product Type P2Y2
Quantity 100 μg
Regulatory Status RUO - research use only