missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ P2X3 Control Peptide
Spécifications
Spécifications
| Gene ID (Entrez) | NM_002559.2 |
| Concentration | 2 mg/mL |
| Content And Storage | -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. |
| Format | Liquid |
| Formulation | Liquid. |
| Gene Symbol | P2RX3, P2X3, MGC129956 |
| Product Type | P2X3, control peptide for AB5895 and AB5896 |
| Quantity | 100 μg |
| Regulatory Status | RUO - research use only |