missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Osteocalcin, Bovine Bone, Protein
Description
Single chain vitamin K-dependent protein produced by osteoblasts. Osteocalcin purified from bovine bone. Binds to phospholipid vesicles in the presence of Ca2+ (Kd = 6μM). Also binds hydroxylapatite. A good marker of bone turnover. May play a role in bone mineralization, bone resorption, and bone formation.
Specifications
Specifications
| Formulation | In 75mM NaCl, 10mM sodium phosphate buffer, pH 7.4. |
| Purification Method | SDS-PAGE |
| Quantity | 100 μg |
| Immunogen | Osteocalcin |
| Storage Requirements | Following initial thaw, aliquot and freeze (−20°C). |
| Regulatory Status | RUO |
| Species | Bovine |
| Recombinant | Native |
| Form | Liquid |
| Purity or Quality Grade | ≥98% |