missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Orexin-2 Receptor Control Peptide

Catalog No. AG794MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AG794MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AG794MI Supplier MilliporeSigma™ Supplier No. AG794

Specifications

Gene ID (Entrez) NM_001526.2
Concentration 1 mg/mL
Content And Storage -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Format Liquid
Formulation Liquid in PBS, pH 7.5 containing 0.05% sodium azide.
Gene Symbol HCRTR2, OTTHUMP00000039969, Ox2r, OX2R
Product Type Orexin-2 Receptor, control peptide for AB3094
Quantity 100 μg
Regulatory Status RUO - research use only