missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Opioid Receptor δ Control Peptide

Catalog No. AG334MI
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AG334MI 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AG334MI Supplier MilliporeSigma™ Supplier No. AG334
Only null left

Specifications

Gene ID (Entrez) NM_000911.3
Content And Storage -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Format Lyophilized
Formulation Lyophilized from PBS, pH 7.4. Reconstitute with 20μL of sterile distilled water.
Gene Symbol OPRD1, OPRD, DOR-1
Product Type Opioid Receptor, δ 358-372, control peptide for AB5505
Quantity 20 μg
Regulatory Status RUO - research use only