missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Normal Human Small Airway Epithelial Cells

Catalog No. SCCE009
Change view
Click to view available options
Quantity:
500,000 cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCCE009 500,000 cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCCE009 Supplier MilliporeSigma™ Supplier No. SCCE009

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

an ideal serum-free culture model for the accurate studies of toxicity, cystic fibrosis, asthma, pathogenesis, pharmacology or airway wound healing.

Millipore’s Normal Human Small Airway Epithelial Cell line, when grown in our recommended medium, provides an ideal serum-free culture model for the accurate studies of toxicity, cystic fibrosis, asthma, pathogenesis, pharmacology or airway wound healing. These cells are not exposed to antimicrobials or phenol red when cultured in EpiGRO™ SAE Complete Media Kit. Normal Human Small Airway Epithelial Cells are cryopreserved at passage 1 to ensure highest viability and plating efficiency.

Specifications

For Use With (Application) Cell Culture
Quantity 500,000 cells
Source Human Small Airway Epithelia