missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ NMDAR1, Control Peptide for 06-314

Catalog No. AG317MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AG317MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AG317MI Supplier MilliporeSigma™ Supplier No. AG317
Only null left

Specifications

Gene ID (Entrez) NM_014839.3
Concentration 1.0 mg/mL
Content And Storage -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Format Liquid
Formulation In PBS
Gene Symbol LPPR4, KIAA0455, PRG1, RP4-788L13.1, PHP1, hCG_2039366, PRG-1
Product Type NMDAR1, control peptide for 06-314
Quantity 100 μg
Regulatory Status RUO - research use only