missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ N27 Rat Dopaminergic Neural Cell Line

Catalog No. SCC048
Change view
Click to view available options
Quantity:
1 x 106Cells/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCC048 1 x 106Cells/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC048 Supplier MilliporeSigma™ Supplier No. SCC048

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

characterized in studies of dopamine biosynthesis, neurotoxicity and used as a dopaminergic neuron model for in vitro and in vivo studies.

Specifications

Content And Storage Store in liquid nitrogen
Tissue E12 Rat Mesencephalic Tissue
For Use With (Application) Stem cell culture and cell culture
Quantity 1 x 106Cells/Vial