missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MultiScreen™HTS 8-Well Strips with Immobilon™-P PVDF Membrane
Description
Includes
Twelve 8-well strips in a support frame. Additional support frames also available separately.
- For ELISpot and protein binding assays
Specifications
Specifications
| Format | Strip Plate |
| Array | 1 x 8 |
| Color | Clear |
| Product Type | Microplate Strip Support Frame |
| For Use With (Equipment) | 8-well strips |
| Material | ABS Plastic |
| No. of Wells | 8 |
| Sterility | Sterile |