missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MultiScreen™ Sealing Tape

Catalog No. MATAHCL00
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
MATAHCL00 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MATAHCL00 Supplier MilliporeSigma™ Supplier No. MATAHCL00

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Choose from opaque or clear

  • Opaque sealing tape is used to seal 100μL plate bottoms read in TopCount™ instruments
  • Clear sealing tape can be used to seal all manufacturers' 300μL plate tops and plate bottoms for MicroBeta™ readers
  • Precut to size

Specifications

Color Clear
Sterility Non-sterile
No. per Pack 100
For Use With (Application) Used to seal all manufacturers 300μL plate tops and plate bottoms for MicroBeta™ readers
Product Type Sealing Tape