missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MultiScreen™ 96-Well, Non-Treated, Flat-Bottom Microplate

Catalog No. MAMCS9610
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
MAMCS9610 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAMCS9610 Supplier MilliporeSigma™ Supplier No. MAMCS9610

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Color:Clear; Sterility:Sterile; Lid:Without Lid

For basolateral feeding and/or co-culture in a single well or 96-well format

  • Use with sterilized MultiScreen filter plates
  • Polystyrene with cut-off corners
  • Sterilized and individually wrapped
  • Untreated surfaces
Single-Well Tray
  • Holds medium and commonly feeds all 96 wells
  • 96-Well Tray
    • Independent wells fit the 96 wells of the membrane plate
    • Patented “tear drop” shaped wells prevent air entrapment under membrane

    Specifications

    Lid Without Lid
    Array 8 x 12
    Color Clear
    Material Polystyrene
    No. of Wells 96
    Sterility Sterile
    Surface Treatment Non-treated
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Cell transport assays, use with sterile MultiScreen filtration plate
    Quantity 10/Pk.