Learn More
MilliporeSigma™ MQuant™ Sulphite Test Strips
To check for presence or absence of residues of Sulphite
Supplier: MilliporeSigma™ 1100130001
Includes: Tube containing 100 strips
Description
To check for presence or absence of residues of Sulphite
- Small and easy to handle
- Flexible and universal application
- Nitroprusside/Zn-hexacyanoferrate reaction method ideal for screening
- Immediate result produced through on-site analysis
- PET film and low reagent content
- Easily disposable
- Color-scale graduation: 10 - 40 - 80 - 180 - 400 mg/L
Reclose the tube containing the test strips immediately after use.
For food and water analysis
Specifications
| Food analysis, particularly in the testing of wine and fruit juices, sulfurated foods, fish shellfish, crustaceans and minced meat products | |
| 10-40-80-180-400mg/L | |
| 10 to 400mg/L | |
| Nitroprusside/Zn-hexacyanoferrate |
| Colorimeter | |
| 100 | |
| 2°C to 8°C | |
| Test Strip |