missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MQuant™ Phosphate Test Strips

Catalog No. M1104280001
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1104280001 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1104280001 Supplier MilliporeSigma™ Supplier No. 1.10428.0001
Only null left

Includes

Reagent

High-tech, fast and simple means for semiquantitative testing

Uses a phosphoromolybdenum blue method.

Small and easy to handle for on-site use
Fast results
Ideal for screening
PET film and low reagent content for easier disposal

Graduations: 10-25-50-100-250-500mg/L

Order Info

No. of tests: 100

Specifications

For Use With (Application) For determining orthophosphate in wastewater, soil samples, fertilizers and food
For Use With (Equipment) Colorimetric
Graduations 10-25-50-100-250-500mg/L
Includes Reagents
Quantity 100 Tests
Range 10 to 500 mg/L
Storage Requirements 15°C to 25°C
Test Method Phosphormolybdenum blue
Type Phosphate Test Strip