missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MQuant™ Peracetic Acid Test Strips

Catalog No. M1179220001
Change view
Click to view available options
Range:
100 to 500mg/L
5 to 50mg/L
500 to 2000mg/L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
M1179220001 500 to 2000mg/L
M1100010001 100 to 500mg/L
M1100840001 5 to 50mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1179220001 Supplier MilliporeSigma™ Supplier No. 1.17922.0001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Tube containing 100 strips

To check for presence of and absence of residues of Peracetic acid

  • Small and easy to handle
  • Flexible and universal application
  • Redox reaction method ideal for screening
  • Immediate result produced through on-site analysis
  • PET film and low reagent content
  • Easily disposable

1.10001.0001
Color-scale graduation: 100 - 150 - 200 - 250 - 300 - 400 - 500mg/L peracetic acid

1.10084.0001
Color-scale graduation: 5 - 10 - 20 - 30 - 50mg/L peracetic acid

Reclose the tube containing the test strips immediately after use.

For food and water analysis

Specifications

For Use With (Application) For food and water analysis
For Use With (Equipment) Colorimeter
Graduations 500-1000-1500-2000mg/L
No. of Tests 100
Range 500 to 2000mg/L
Storage Requirements 2°C to 8°C
Test Method Redox Reaction
Type Test Strip