missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MQuant™ Nitrite Test Strips

To determine nitrite in analysis of water and cooling lubricants

Supplier:  MilliporeSigma™ 1100070002

 View more versions of this product

Catalog No. M1100070002

Only null left

Includes: Tube containing 100 strips

Description

Description

  • Small and easy to handle
  • Flexible and universal application
  • Griess' reaction method ideal for screening
  • Immediate result produced through on-site analysis
  • PET film and low reagent content
  • Easily disposable

1.10007.0001
Color-scale graduation: 2 - 5 - 10 - 20 - 40 - 80mg/L

1.10022.0001
Color-scale graduation: 0.1 - 0.3 - 0.6 - 1 - 2 - 3g/L

Reclose the tube containing the test strips immediately after use.

For food and water analysis

Specifications

Specifications

To determine nitrite in analysis of water and cooling lubricants
2-5-10-20-40-80mg/L
2 to 80mg/L NO2-
Griess' reaction
Colorimeter
25
2°C to 8°C
Test Strip
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions