missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MQuant™ Nitrate Test Strips
To determine nitrate in aqueous media and food
$276.39 - $2,074.24
Specifications
| For Use With (Application) | For food and water analysis |
|---|---|
| For Use With (Equipment) | Colorimeter |
| Graduations | 10-25-50-100-250-500mg/L |
| Storage Requirements | 2°C to 8°C |
| Test Method | Modified Griess' reaction |
Includes: Tube containing 100 strips
| Catalog Number | Mfr. No. | No. of Tests | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Tests | Price | Quantity & Availability | |||||
|
M1100200001
|
MilliporeSigma™
1.10020.0001 |
100 |
Each for $276.39
|
|
|||||
|
M1100920021
|
MilliporeSigma™
1.10092.0021 |
1000 |
Each for $2,074.24
|
|
|||||
Description
- Small and easy to handle
- Flexible and universal application
- Modified Griess' reaction method ideal for screening
- Immediate result produced through on-site analysis
- PET film and low reagent content
- Easily disposable
- Color-scale graduation: 10 - 25 - 50 - 100 - 250 - 500mg/L
Reclose the tube containing the test strips immediately after use.
For food and water analysis
Specifications
| For food and water analysis | |
| 10-25-50-100-250-500mg/L | |
| Modified Griess' reaction |
| Colorimeter | |
| 2°C to 8°C | |
| Test Strip |