Learn More
MilliporeSigma™ MQuant™ Cobalt Test Strips
To check for the presence or absence of residues of cobalt
Supplier:  MilliporeSigma™ 1100020001
Catalog No. M1100020001
Only null left
Includes: Tube containing 100 strips
Description
Description
  • Small and easy to handle
  • Flexible and universal application
  • Rhodanide method ideal for screening
  • Immediate result produced through on-site analysis
  • PET film and low reagent content
  • Easily disposable
  • Color-scale graduation: 10 - 30 - 100 - 300 - 1000mg/L
Testing of waste water, electroplating baths, in electronics industry, metal industry, in assaying and refining of ores
Specifications
Specifications
Testing of wastewater, electroplating baths, in electronics industry, metal industry, in assaying and refining of ores
10-30-100-300-1000mg/L
10 to 1000mg/L
Rhodanide
Colorimeter
100
15°C to 25°C
Test Strip
Product Suggestions
Product Suggestions