missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MQuant™ Chromate Test Strips
Description
Includes
Reagent
Uses diphenylcarbizide method.
- Small and easy to handle for on-site use
- Fast results
- Ideal for screening
- PET film and low reagent content for easier disposal
- Graduations: 3-10-30-100mg/L
Specifications
Specifications
| For Use With (Application) | The chromate test can be used to monitor wastewater discharged from electroplating and pickling plants, in cement and for rapidly and reliably determining of chromium(VI) salts |
| Graduations | 3-10-30-100mg/L |
| Includes | 1 bottle of reagent CrO4-1, 1 test vessel |
| Quantity | Each |
| Range | 3 to 100 mg/L |
| Storage Requirements | 15°C to 25°C |
| Test Method | Diphenylcarbazide |
| Type | MQuant™ Chromate Test Strip |