missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MQuant™ Chlorine Test Strips
Description
Includes
Tube containing 75 strips
- Small and easy to handle
- Flexible and universal application
- Redox reaction method ideal for screening
- Immediate result produced through on-site analysis
- PET film and low reagent content
- Easily disposable
- Color-scale graduation: 0.5 - 1 - 2 - 5 - 10 - 20mg/L
Reclose the tube containing the test strips immediately after use
For water analysis
Specifications
Specifications
| For Use With (Application) | To check for presence of chlorine in disinfectants and water |
| For Use With (Equipment) | Colorimeter |
| Graduations | 0.5-1-2-5-10-20mg/L |
| No. of Tests | 75 |
| Range | 0.5 to 20mg/L |
| Storage Requirements | 2°C to 8°C |
| Test Method | Redox Reaction |
| Type | Test Strip |