missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MQuant™ Chlorine Test Strips

Catalog No. M1179250001
Change view
Click to view available options
Range:
0.5 to 20mg/L
25 to 500mg/L
No. of Tests:
100
75
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range No. of Tests
M1179250001 0.5 to 20mg/L 75
M1179240001 25 to 500mg/L 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1179250001 Supplier MilliporeSigma™ Supplier No. 1.17925.0001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Tube containing 75 strips

To determine the presence of residues of chlorine

  • Small and easy to handle
  • Flexible and universal application
  • Redox reaction method ideal for screening
  • Immediate result produced through on-site analysis
  • PET film and low reagent content
  • Easily disposable
  • Color-scale graduation: 0.5 - 1 - 2 - 5 - 10 - 20mg/L

Reclose the tube containing the test strips immediately after use

For water analysis

Specifications

For Use With (Application) To check for presence of chlorine in disinfectants and water
For Use With (Equipment) Colorimeter
Graduations 0.5-1-2-5-10-20mg/L
No. of Tests 75
Range 0.5 to 20mg/L
Storage Requirements 2°C to 8°C
Test Method Redox Reaction
Type Test Strip