missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MQuant™ Carbonate Hardness Test Strips
Description
Includes
Total Hardness Test (100 visual test strips), measuring range >6 - >31°e
- Small and easy to handle
- Flexible and universal application
- Mixed indicator method ideal for screening
- Immediate result produced through on-site analysis
- PET film and low reagent content
- Easily disposable
- Color-scale graduation: 5 - 10 - 15 - 20 - 30°e
Reclose the tube containing the test strips immediately after use.
Specifications
Specifications
| For Use With (Application) | Rapid determination of carbonate hardness |
| For Use With (Equipment) | Colorimeter |
| Graduations | 5 to 30°e |
| No. of Tests | 100 |
| Range | 5 to 30°e |
| Storage Requirements | 15°C to 25°C |
| Test Method | Mixed Indicator |
| Type | Test Strip |