missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MQuant™ Calcium Test Kit

Catalog No. M1100830001
Change view
Click to view available options
Range:
10 to 100mg/L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
M1100830001 10 to 100mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1100830001 Supplier MilliporeSigma™ Supplier No. 1.10083.0001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Reagent

High-tech, fast and simple means for semiquantitative testing

Uses a Glyoxal bishydroxyanil method.

  • Small and easy to handle for on-site use
  • Fast results
  • Ideal for screening
  • PET film and low reagent content for easier disposal

Graduations: 10-25-50-100mg/L

Order Info

Pack of 60 tests

Specifications

For Use With (Application) For rapid determination of calcium in aqueous media
For Use With (Equipment) Colorimetric
Graduations 10-25-50-100mg/L
Includes 1 bottle reagent Ca-1, 1 bottle reagent Ca-2, 1 test vessel
Quantity 60 Tests
Range 10 to 100mg/L
Storage Requirements 2°C to 8°C
Test Method Glyoxal bishydroxyanil
Type Calcium Test Strip