Learn More
MilliporeSigma™ MQuant™ Blank Test Strips
Description
- Ideal to test if sample solutions turn the blank a different color — potenitally affecting results of other MQuant strips
- Small and easy to handle for on-site use
- Fast results
- Ideal for screening
- PET film and low reagent content for easier disposal
Specifications
Specifications
| For Use With (Application) | For the semiquantitative detection of ions and of inorganic and organic substances and can easily be used anywhere |
| Quantity | Each |
| Type | Blank Test Strip |