missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MQuant™ Arsenic Test Strips
Description
Includes
Reagent
Comfortable and secure transport for on-site analyses.
- Designed for upright transport of reagents
- Solid acid is used as a reagent to avoid transporting liquid acid and zinc
- Two reagent bottles to conduct two tests simultaneously
- Modified Gutzeit test
- PET film and low reagent content for easier disposal
- Small and easy to handle for on-site use
- Fast results
- Ideal for screening
- Available in practical case (M1179270001) or stable plastic box (M1179170001)
- M1179170001 Graduations: 0.02-0.05-0.1-0.2-0.5mg/L; 0.1-0.5-1.0-1.7-3mg/L
- M1179270001 Graduations: 0.005-0.01-0.025-0.05-0.1-0.25-0.5mg/L
Specifications
Specifications
| For Use With (Application) | For measuring arsenic in water, soil extracts, pharmaceutical products, prepared biological specimens and liquid foods |
| Graduations | 0.02-0.05-0.1-0.2-0.5mg/L |
| Includes | 2 bottles of reagent As-1, 1 bottle of reagent As-2, 1 graduated 5-ml plastic syringe, 1 red dosing spoon, 2 reaction tubes with screw caps |
| Quantity | 100 Tests |
| Range | 0.02 to 3 mg/L |
| Storage Requirements | 15°C to 25°C |
| Test Method | Modified Gulzeit test |
| Type | MQuant™ Arsenic Test Strip |