Learn More
MilliporeSigma™ Millicell™ EZ Slides
Culture, fix, stain, and view cells all in one device
Supplier:  MilliporeSigma™ PEZGS0416
 View more versions of this product
Catalog No. PEZGS0416
Only null left
Explore available promotions
Description
Description
  • Polypropylene wells are resistant to most fixatives—cells may be fixed and stained with wells still attached
  • Breakable tabs for easy well removal—no tools required to remove wells from slide
  • No adhesive used to attach wells to slide
  • No leaky wells
  • Raised corner ridges on lid facilitate stacking of multiple units in the incubator
  • Noncytotoxic and nonpyrogenic
  • Sterile
  • Slide holder (available separately) protects slide from damage
Specifications
Specifications
EZ Slide
Clear
1.7 cm2/well
Glass
Simplify your cell analysis by using the Millicell EZ SLIDE glass slide to culture, fix, stain and view your sample all in one device
4
Polypropylene
Polystyrene
0.5 to 1.7 mL/Well
Sterile
Product Suggestions
Product Suggestions