Learn More
MilliporeSigma™ Millicell™ EZ Slides
Catalog No. PEZGS0816
Change view
Click to view available options
Wells:
4
8
Quantity:
16 Pk.
96 Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells Quantity
PEZGS0816 8 16 Pk.
PEZGS0416 4 16 Pk.
PEZGS0496 4 96 Pk.
PEZGS0896 8 96 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PEZGS0816 Supplier MilliporeSigma™ Supplier No. PEZGS0816
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Culture, fix, stain, and view cells all in one device
  • Polypropylene wells are resistant to most fixatives—cells may be fixed and stained with wells still attached
  • Breakable tabs for easy well removal—no tools required to remove wells from slide
  • No adhesive used to attach wells to slide
  • No leaky wells
  • Raised corner ridges on lid facilitate stacking of multiple units in the incubator
  • Noncytotoxic and nonpyrogenic
  • Sterile
  • Slide holder (available separately) protects slide from damage
Specifications
Type EZ Slide
Wells 8
Color Clear
Well Material Polypropylene
Culture Area 0.7 cm2/well
Lid Material Polystyrene
Material Glass
Volume (Metric) Working 0.2 to 0.7 mL/Well
For Use With (Application) Simplify your cell analysis by using the Millicell EZ SLIDE glass slide to culture, fix, stain and view your sample all in one device
Sterility Sterile
Quantity 16 Pk.
Show More Show Less