missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Milli-Q™ Ultrapure Water Systems Accessory, Cartridges
Description
Choose from a wide selection of replacement cartridges
Filter Types
- Activated carbon
- Ion exchange
- Membrane
Specifications
Specifications
| Purification Method | Ion Exchange |
| For Use With (Equipment) | Milli-Q Systems |
| Model | Quantum ICP |
| Type | Polishing Cartridge |